missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Soda Lime Glass Threaded Bottles

Product Code. 10223312 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2500 mL
Coating:
None
ethylene-acrylate coating
Unit Size:
Each
Product Code. Capacity (Metric) Coating unitSize
10223312 500 mL None Each
10407231 2500 mL ethylene-acrylate coating Each
10400602 100 mL None Each
10723273 250 mL None Each
10233312 2500 mL None Each
10660953 1000 mL None Each
10390092 100 mL ethylene-acrylate coating Each
7 options
This item is not returnable. View return policy

Product Code. 10223312

Brand: BRAND™ 704016

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Soda-lime glass bottle features ethylene-acrylate coating that envelops the glass bottle like a protection skin.

  • Soda-lime glass (amber)
  • Screw-cap, polypropene (PP), pouring ring PE-LD
  • Round glass narrow mouth bottles
  • Space-saving square base
  • Available ethylene-acrylate-coated or uncoated
  • The coating envelops the glass bottle like a protection skin
  • Should the bottle break, the splintering hazards are reduced considerably
  • The maximum operating temperature for coated bottles is 80 °C
  • To preserve the coating, do not clean at temperatures exceeding 60 ºC
  • With PP screw cap GL45
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Coating None
Color Amber
Material Soda Lime Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.