missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Round glass narrow mouth bottles
- Space-saving square base
- Available ethylene-acrylate-coated or uncoated
- The coating envelops the glass bottle like a protection skin
- Should the bottle break, the splintering hazards are reduced considerably
- The maximum operating temperature for coated bottles is 80 °C
- To preserve the coating, do not clean at temperatures exceeding 60 ºC
- With PP screw cap GL45
Specifications
Specifications
| Capacity (Metric) | 2500 mL |
| Coating | ethylene-acrylate coating |
| Color | Amber |
| Material | Soda Lime Glass |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?