missing translation for 'onlineSavingsMsg'
Learn More

Soda lime, pure, granular, without indicator, Thermo Scientific™

Product Code. 17525595 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17525595 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17525595 Supplier Thermo Scientific Acros Supplier No. 472160010
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 8006-28-8
Color White to beige to light gray
Physical Form Granules
Quantity 1 kg
Loss on Drying ≤20 % (10 g, 200°C, 2 h)
Packaging Glass Bottle
Molecular Formula CaHNaO2
MDL Number 00134094
Merck Index 13-8640
Solubility Information Slightly soluble in water
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES [OH-].[Na+].O=[Ca]
IUPAC Name sodium oxocalcium hydroxide
Molecular Weight (g/mol) 96.07
Grade Ungraded
Chemical Name or Material Soda lime
Show More Show Less
compliance-icons
Product Identifier
  • Soda lime, without indicator
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Soda lime, Calcium hydroxide, Sodium hydroxide, Potassium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.