Learn More
Sodium 1-dodecanesulfonate, 99%
Description
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 10 g |
| Beilstein | 3919536 |
| Solubility Information | Soluble in water. |
| Formula Weight | 272.39 |
| Percent Purity | 99% |
| Chemical Name or Material | Sodium 1-dodecanesulfonate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.