missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%

Product Code. 11464303
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464303 100 g 100g
11444303 5 g 5g
11454303 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464303 Supplier Thermo Scientific Alfa Aesar Supplier No. A10917.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22767-50-6 | C7H15NaO3S | 202.244 g/mol

HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
MDL Number MFCD00007543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Odor Odorless
Quantity 100 g
Beilstein 3731762
Solubility Information Soluble in water.
Formula Weight 202.24
Percent Purity 99%
Assay (dry wt.)
Chemical Name or Material Sodium 1-heptanesulfonate, Water <2%
compliance-icons
Product Identifier
  • Sodium 1-heptanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Heptanesulfonic acid, sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.