missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-hexanesulfonate, 99%

Product Code. 11440833
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11440833

Brand: Thermo Scientific Alfa Aesar A13278.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol

Sodium 1-hexanesulfonate is used as a base in the organic coupling reactions such as Suzuki coupling reactions. It is used as an ion pairing reagent for HPLC and involved in the analysis of peptides and proteins. It is also used in high performance capillary electrophoresis analysis of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-hexanesulfonate is used as a base in the organic coupling reactions such as Suzuki coupling reactions. It is used as an ion pairing reagent for HPLC and involved in the analysis of peptides and proteins. It is also used in high performance capillary electrophoresis analysis of peptides.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium;hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Assay Percent Range 99%
Quantity 5 g
Beilstein 3727014
Solubility Information Soluble in water.
Formula Weight 188.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Sodium 1-hexanesulfonate
compliance-icons
Product Identifier
  • Sodium 1-hexanesulfonate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.