missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%

Product Code. 11375337
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375337 5 g 5g
11355337 25 g 25g
11365337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375337 Supplier Thermo Scientific Alfa Aesar Supplier No. 042636.06
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Ion-pair reagent for HPLC analysis | CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol

Ion-pair reagent for HPLC analysisSodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium;octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Spezifikation

Melting Point >300°C
Quantity 5 g
Solubility Information Soluble in water producing a clear and colorless solution.
Formula Weight 216.28 (Anhydrous)
Percent Purity ≥99%
Grade HPLC
Physical Form Crystalline
Chemical Name or Material Sodium 1-octanesulfonate hydrate
compliance-icons
Produktidentifikator
  • Sodium 1-octanesulfonate hydrate, HPLC grade
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Sodium 1-octanesulfonate hydrate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.