missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-pentanesulfonate, 99% (dry wt.), water <1.5%

Product Code. 11405546
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405546 25 g 25g
11495536 5 g 5g
11415546 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405546 Supplier Thermo Scientific Alfa Aesar Supplier No. B20699.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22767-49-3 | C5H11NaO3S | 174.19 g/mol

Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007541,MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
PubChem CID 23664617
IUPAC Name sodium;pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 3725147
Solubility Information Soluble in water.
Formula Weight 174.19
Percent Purity 99%
Assay (dry wt.), water <1%
Chemical Name or Material Sodium 1-pentanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.