missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Propanethiolate (Technical Grade), TRC

Product Code. 30413746
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413746 Supplier Toronto Research Chemicals Supplier No. TRCS6732801G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 1-Propanethiolate (Technical Grade)
CAS 6898-84-6
Molecular Formula C3H7NaS
Quantity 1 g
Synonym 1-Propanethiol','Sodium Salt, Propylmercaptan Sodium Salt, Sodium 1-Propanethiolate, Sodium n-Propylthiolate, Sodium Propanethiolate, Sodium Propyl Sulfide, Sodium Propylmercaptide
InChI Formula InChI=1S/C3H8S.Na/c1-2-3-4;/h4H,2-3H2,1H3;/q;+1/p-1
SMILES [Na+].CCC[S-]
IUPAC Name sodium;propane-1-thiolate
Molecular Weight (g/mol) 98.14
Formula Weight 98.0166
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.