missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,4-pentanedionate hydrate, 94%

Product Code. 11466916
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466916 50 g 50g
11476916 250 g 250g
11456916 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466916 Supplier Thermo Scientific Alfa Aesar Supplier No. B24009.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 86891-03-4 | C5H7NaO2 | 122.10 g/mol

Sodium 2,4-pentanedionate hydrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2,4-pentanedionate hydrate is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, water/ moisture, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86891-03-4
Molecular Formula C5H7NaO2
Molecular Weight (g/mol) 122.10
MDL Number MFCD00078034
InChI Key AIWZOHBYSFSQGV-UHFFFAOYSA-M
Synonym sodium 2,4-pentanedionate, sodium 2e-4-oxopent-2-en-2-olate hydrate
PubChem CID 90476915
IUPAC Name sodium;(E)-4-oxopent-2-en-2-olate;hydrate
SMILES [Na+].CC([O-])=CC(C)=O

Specifications

Melting Point 228°C to 230°C
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 122.11 (Anhydrous)
Percent Purity 95%
Chemical Name or Material Sodium 2,4-pentanedionate hydrate
compliance-icons
Product Identifier
  • Sodium 2,4-pentanedionate hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.