missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Hydroxybutyrate, TRC

Product Code. 30421072
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421072 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421072 Supplier Toronto Research Chemicals Supplier No. TRCS5094251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 2-Hydroxybutyrate
CAS 5094-24-6
Molecular Formula C4 H7 O3 . Na
Quantity 1 g
Synonym Sodium α-Hydroxybutyrate, DL-α-Hydroxybutyric Acid Sodium Salt, Sodium (DL)-α-Hydroxybutyrate, Sodium (±)-2-Hydroxybutyrate, Sodium 2-Hydroxybutanoate, 2-Hydroxybutanoic Acid Sodium Salt, 2-Hydroxybutyric Acid Sodium Salt
InChI Formula InChI=1S/C4H8O3.Na/c1-2-3(5)4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1
SMILES [Na+].CCC(O)C(=O)[O-]
IUPAC Name sodium;2-hydroxybutanoate
Molecular Weight (g/mol) 126.086
Formula Weight 126.0293
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.