missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3,5-dicarboxybenzenesulfonate, 97%, BLD Pharm™

Product Code. 31108010
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31108010 100 g
31104794 25 g
31106460 1 kg
31096530 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31108010 Supplier BLD Pharm Supplier No. BD107381100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6362-79-4
Molecular Formula C8H5NaO7S
MDL Number MFCD00007495
Quantity 100 g
SMILES O=S(C1=CC(C(O)=O)=CC(C(O)=O)=C1)([O-])=O.[Na+]
Molecular Weight (g/mol) 268.18
Percent Purity 97%
Chemical Name or Material Sodium 3,5-dicarboxybenzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.