missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-bromopropanesulfonate, 97%

Product Code. 11468777
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11468777

Brand: Thermo Scientific Alfa Aesar A19930.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55788-44-8 | C3H7BrNaO3S | 226.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55788-44-8
Molecular Formula C3H7BrNaO3S
Molecular Weight (g/mol) 226.04
MDL Number MFCD00024999
InChI Key ONPNLMMBTJZJRQ-UHFFFAOYSA-N
Synonym sodium 3-bromopropanesulfonate, sodium 3-bromopropane-1-sulfonate, 3-bromopropanesulfonic acid sodium salt, acmc-209lpd, sodium 3-bromo-1-propanesulfonate, sodium 3-bromo-propane-1-sulphonate, 3-bromopropane sulfonic acid sodium salt, 3-bromo-1-propanesulfonic acid sodium salt, 1-propanesulfonic acid, 3-bromo-, sodium salt
PubChem CID 23679056
IUPAC Name sodium;3-bromopropane-1-sulfonate
SMILES [Na].OS(=O)(=O)CCCBr

Specifications

Melting Point ∼256°C (decomposition)
Quantity 100 g
Formula Weight 225.05
Percent Purity 97%
Chemical Name or Material Sodium 3-bromopropanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.