missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-nitrobenzenesulfonate, 98%, water <2%

Product Code. 11473943
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473943 100 g 100g
11483943 500 g 500g
11493943 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473943 Supplier Thermo Scientific Alfa Aesar Supplier No. A10244.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-68-4 | C6H4NNaO5S | 225.15 g/mol

Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

Solubility
Soluble in water 200 g/L (20C).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-68-4
Molecular Formula C6H4NNaO5S
Molecular Weight (g/mol) 225.15
MDL Number MFCD00007490
InChI Key LJRGBERXYNQPJI-UHFFFAOYSA-M
Synonym sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
PubChem CID 31389
IUPAC Name sodium;3-nitrobenzenesulfonate
SMILES C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

Melting Point 350°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water 200g/L (20C).
Formula Weight 225.16 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium 3-nitrobenzenesulfonate, Water <2%
compliance-icons
Product Identifier
  • Sodium 3-nitrobenzenesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium 3-nitrobenzenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.