missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), BLD Pharm™

Product Code. 31099684
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31099684 25 g
31101041 5 g
31095468 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31099684 Supplier BLD Pharm Supplier No. BD15053125g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 632-69-9
Molecular Formula C20H2Cl4I4Na2O5
MDL Number MFCD00151169
Quantity 25 g
SMILES O=C1OC2(C3=C(OC4=C2C=C(I)C([O-])=C4I)C(I)=C([O-])C(I)=C3)C5=C1C(Cl)=C(Cl)C(Cl)=C5Cl.[Na+].[Na+]
Molecular Weight (g/mol) 1017.64
Chemical Name or Material Sodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.