missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Acetylbenzenesulfonate, TRC

Product Code. 30420309
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420309 500 mg -
30421845 250 mg -
30414993 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420309 Supplier Toronto Research Chemicals Supplier No. TRCS644833500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61827-67-6
Molecular Formula C8H7NaO4S
Molecular Weight (g/mol) 222.19
Synonym 4-Acetylbenzenesulfonic Acid Sodium Salt, p-Acetylbenzenesulfonic Acid Sodium Salt
IUPAC Name sodium;4-acetylbenzenesulfonate
SMILES [Na+].CC(=O)c1ccc(cc1)S(=O)(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H8O4S.Na/c1-6(9)7-2-4-8(5-3-7)13(10,11)12;/h2-5H,1H3,(H,10,11,12);/q;+1/p-1
Formula Weight 221.9963
Chemical Name or Material Sodium 4-Acetylbenzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.