missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Acetylbenzenesulfonate, TRC

Product Code. 30421845
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421845 250 mg -
30420309 500 mg -
30414993 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421845 Supplier Toronto Research Chemicals Supplier No. TRCS644833250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61827-67-6
Molecular Formula C8H7NaO4S
Molecular Weight (g/mol) 222.19
Synonym 4-Acetylbenzenesulfonic Acid Sodium Salt, p-Acetylbenzenesulfonic Acid Sodium Salt
IUPAC Name sodium;4-acetylbenzenesulfonate
SMILES [Na+].CC(=O)c1ccc(cc1)S(=O)(=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H8O4S.Na/c1-6(9)7-2-4-8(5-3-7)13(10,11)12;/h2-5H,1H3,(H,10,11,12);/q;+1/p-1
Formula Weight 221.9963
Chemical Name or Material Sodium 4-Acetylbenzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.