missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-ethynylbenzoate, 97%

Product Code. 11384176
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384176 1 g 1g
11304186 250 mg 250mg
11394176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384176 Supplier Thermo Scientific Alfa Aesar Supplier No. L19292.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 144693-65-2 | C9H5NaO2 | 168.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144693-65-2
Molecular Formula C9H5NaO2
Molecular Weight (g/mol) 168.127
MDL Number MFCD03426026
InChI Key VXBLAFDQGACQAR-UHFFFAOYSA-M
Synonym sodium 4-ethynylbenzoate, 4-ethynylbenzoic acid sodium salt, 4-ethynyl-benzoic acid sodium salt, benzoic acid, 4-ethynyl-, sodium salt, benzoic acid,4-ethynyl-, sodium salt 1:1, 4-ethynyl benzoic acid sodium salt, sodium p-ethynylbenzoate, acmc-20egl3, 4-ethynylbenzoic acid sodium, 4-ethynylbenzoic-acid sodium salt
PubChem CID 23720426
IUPAC Name sodium;4-ethynylbenzoate
SMILES C#CC1=CC=C(C=C1)C(=O)[O-].[Na+]

Specifications

Quantity 1 g
Beilstein 5836538
Formula Weight 168.13
Percent Purity 97%
Chemical Name or Material Sodium 4-ethynylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.