missing translation for 'onlineSavingsMsg'
Learn More

Sodium 6-chloropyridazine-3-carboxylate, Thermo Scientific™

Product Code. 10420870
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10420870 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10420870 Supplier Thermo Scientific Maybridge Supplier No. MAY00098DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 89799-77-9
Packaging Amber Glass Bottle
Molecular Formula C5H2ClN2NaO2
Quantity 1 g
Synonym sodium 6-chloropyridazine-3-carboxylate, c5h2cln2o2.na, sodium 6-chloro-pyridazine-3-carboxylate, 6-chloropyridazine-3-carboxylic acid, sodium salt, 3-pyridazinecarboxylic acid, 6-chloro-, sodium salt
InChI Key SZIZZPDMABOKHD-UHFFFAOYSA-M
SMILES C1=CC(=NN=C1C(=O)[O-])Cl.[Na+]
IUPAC Name sodium;6-chloropyridazine-3-carboxylate
Molecular Weight (g/mol) 180.523
PubChem CID 24820514
Formula Weight 180.53
Percent Purity 97%
Chemical Name or Material Sodium 6-chloropyridazine-3-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.