missing translation for 'onlineSavingsMsg'
Learn More

Sodium, 99.8%, oiled sticks, wrapped in aluminum foil

Product Code. 10207882 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10207882 100 g 100g
10084322 250 g 250g
10757693 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10207882 Supplier Thermo Scientific Acros Supplier No. 390121000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-23-5 | Na | 22.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-23-5 , 8042-47-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Color Gray
Melting Point 98.0°C
Boiling Point 883.0°C
Quantity 100 g
Assay Percent Range 99.8%
Form Stick
Linear Formula Na
Merck Index 15, 8708
Formula Weight 23.99
Percent Purity 99.8%
Chemical Name or Material Sodium
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium, under mineral oil
Signal Word
  • Danger
Hazard Category
  • Substance which, in contact with water, emits flammable gases Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.