missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, anhydrous, 99%

Produktkod. 11444584
Change view
Klicka för att se tillgängliga alternativ
Quantity:
250 g
1000 g
5000 g
Förpackningsstorlek:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
11444584 5000 g 5000g
11434584 250 g 250g
11424584 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11444584 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A13184.0I
 Restricted Item
In Stock
Lägg i varukorgen
Edge
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.

Solubility
Very soluble in water. Soluble in alcohol.

Notes
Hygroscopic. It is incompatible with strong oxidizing agents, halogens and it is moisture sensitive.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifikationer

Melting Point 324°C
Density 1.528
pH 7.5 to 9
Assay Percent Range 99%
Linear Formula NaOOCCH3
Quantity 5000 g
Beilstein 3595639
Sensitivity Hygroscopic
Merck Index 14,8571
Solubility Information Very soluble in water. Soluble in alcohol.
IUPAC Name sodium;acetate
Formula Weight 82.03
Percent Purity 99%
Chemical Name or Material Sodium acetate, anhydrous
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.