missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents

Product Code. 10103243
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10103243 500 g Poly Bottle 500g
10498913 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103243 Supplier Fisher BioReagents Supplier No. BP333500
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Acetate is added to DNA reactions to facilitate ethanol precipitation of the DNA.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324°C
Color White
pH 7.5 to 9.2
Physical Form Powder/Solid
Quantity 500 g
Loss on Drying 1% max. (at 105°C)
Packaging Poly Bottle
Assay Percent Range ≥99.0 %
Linear Formula CH3COONa
Merck Index 15, 8709
IUPAC Name sodium acetate
Formula Weight 82.03
Percent Purity ≥99.0%
Grade Ungraded
Chemical Name or Material Sodium Acetate Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.