missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents

Product Code. 10498913
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10498913

Brand: Fisher BioReagents BP3331

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sodium Acetate is added to DNA reactions to facilitate ethanol precipitation of the DNA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324°C
Color White
pH 7.5 to 9.2
Physical Form Powder/Solid
Quantity 1 kg
Assay Percent Range ≥99.0 %
Linear Formula CH3COONa
Merck Index 15, 8709
IUPAC Name sodium;acetate
Formula Weight 82.03
Percent Purity ≥99.0%
Loss on Drying 1% max. (at 105°C)
Packaging Glass Bottle
Chemical Name or Material Sodium Acetate Anhydrous
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.