Learn More
Sodium Acetate Buffer Solution pH 4.5, Fisher Chemical™
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Brand: Spectracer SOL 037 1L
Code : 04
Additional Details : Weight : 1.00000kg
Specifications
4.5 | |
Buffer | |
Liquid | |
1000mL |