missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10376390 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10376390 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10376390 Supplier JT Baker Supplier No. 0256.1000
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets ACS Specifications

Specifications

Boiling Point 120°C to 123°C (1013 hPa)
Melting Point 58°C
CAS 6131-90-4
pH 7.5 to 9.2 (5% Solution at 25°C)
Quantity 1 kg
Assay Percent Range 99.0 to 101.0%
Molecular Formula C2H9NaO5
MDL Number MFCD00071557
Synonym Acetic acid sodium salt trihydrate
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES O.O.O.[Na+].CC([O-])=O
IUPAC Name sodium acetate trihydrate
Molecular Weight (g/mol) 136.08
Packaging Plastic bottle for solids
Density 1.4 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.