missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, BAKER, J.T. Baker™

Product Code. 10589020 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10589020 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10589020 Supplier JT Baker Supplier No. 0517.1000
  • ONLINE EXCLUSIVE PRICE
    40.47 €
    31.82 € / 1kg
    Save 8.65 €
    21% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

Specifications

CAS 6131-90-4
Quantity 1 kg
Molecular Formula C2H9NaO5
Synonym Acetic acid sodium salt trihydrate
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES O.O.O.[Na+].CC([O-])=O
IUPAC Name sodium acetate trihydrate
Molecular Weight (g/mol) 136.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.