missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Trihydrate, for HPLC, Certified, Fisher Chemical

Product Code. 10733704 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10733704 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10733704 Supplier Fisher Chemical Supplier No. S/2046/60
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 58°C
CAS 6131-90-4
Color White
pH 8 to 9.5
Physical Form Solid
Quantity 1 kg
Molecular Formula C2H9NaO5
MDL Number 71557
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES CC(=O)[O-].O.O.O.[Na+]
IUPAC Name sodium;acetate;trihydrate
Molecular Weight (g/mol) 136.079
PubChem CID 23665404
ChEBI CHEBI:32138
Formula Weight 136.08g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.