missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al, Solstice
BIOPHARMA

Product Code. 15642720 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
25kg
5kg
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15642720 5 kg 5kg
15612720 1 kg 1kg
15622720 2.5 kg 2.5kg
15652720 6 x 1 kg Pack of 6
15662720 6 x 2.5 kg Pack of 6
15632720 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15642720 Supplier Honeywell-Fluka Supplier No. 250225KG
  • ONLINE EXCLUSIVE PRICE
    214.00 €
    192.80 € / 5kg
    Save 21.20 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number MFCD00071557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Physical Form Crystalline Powder
Quantity 5 kg
Linear Formula CH3COONa · 3H2O
UN Number NONH for all modes of transport
Beilstein 3732037
Formula Weight 136.08g/mol
Percent Purity 99 to 101% (calc. to the dried substance)
Grade puriss.
Ignition Point 1112°F
Chemical Name or Material Sodium Acetate Trihydrate, Meets analytical specification of Ph.Eur.BP, USP, FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.