missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Solstice

Product Code. 15693800
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
50 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
500g
50kg
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15693800 1 kg 1kg
15613810 500 g 500g
15603810 2.5 kg 2.5kg
15653810 6 x 500 g Pack of 6
15633810 6 x 1 kg Pack of 6
15643810 6 x 2.5 kg Pack of 6
15623810 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15693800 Supplier Honeywell-Fluka Supplier No. 323181KG
  • ONLINE EXCLUSIVE PRICE
    71.00 €
    52.37 € / 1kg
    Save 18.63 €
    26% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Linear Formula CH3COONa · 3H2O
MDL Number MFCD00071557
UN Number NONH for all modes of transport
Beilstein 3732037
Formula Weight 136.08g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Ignition Point 1112°F
Chemical Name or Material Sodium Acetate Trihydrate, Reag. Ph. Eur. reag. ISO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.