missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Product Code. p-7072069 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
50 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
500g
50kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15613810

Brand: Honeywell-Fluka 32318500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Quantity 500 g
Linear Formula CH3COONa · 3H2O
MDL Number MFCD00071557
UN Number NONH for all modes of transport
Beilstein 3732037
Formula Weight 136.08g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Ignition Point 1112°F
Chemical Name or Material Sodium Acetate Trihydrate, Reag. Ph. Eur. reag. ISO
Product Identifier
  • Sodium acetate trihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.