missing translation for 'onlineSavingsMsg'
Learn More

Sodium amide, 95%, pure

Product Code. 10286250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
100g
2.5kg
500g
Product Code. Quantity Packaging unitSize
10286250 100 g Glass bottle 100g
10654892 500 g Glass bottle 500g
11964391 2.5 kg Plastic Drum 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 10286250

Brand: Thermo Scientific Acros 197051000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-92-5 | H2NNa | 39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-92-5
InChI Key ODZPKZBBUMBTMG-UHFFFAOYSA-N
Synonym sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine
PubChem CID 24533
IUPAC Name sodium;azanide
SMILES [NH2-].[Na+]

Specifications

Boiling Point 400°C
Melting Point 210°C
Color Gray-White to White
Physical Form Powder, Irregular Flakes or Granules
Quantity 100 g
Assay Percent Range 95%
Molecular Formula H2NNa
Linear Formula NaNH2
Fieser 01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288
Merck Index 15,8714
Solubility Information Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol
Molecular Weight (g/mol) 39
Formula Weight 39
Percent Purity 95%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Sodium amide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium amide
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • EUH019-May form explosive peroxides.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P223-Do not allow contact with water.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.