missing translation for 'onlineSavingsMsg'
Learn More

Sodium amide, 95%, pure

Product Code. 10654892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
100g
2.5kg
500g
Product Code. Quantity Packaging unitSize
10654892 500 g Glass bottle 500g
10286250 100 g Glass bottle 100g
11964391 2.5 kg Plastic Drum 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 10654892

Brand: Thermo Scientific Acros 197055000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-92-5 | H2NNa | 39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-92-5
InChI Key ODZPKZBBUMBTMG-UHFFFAOYSA-N
Synonym sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine
PubChem CID 24533
IUPAC Name sodium;azanide
SMILES [NH2-].[Na+]

Specifications

Boiling Point 400°C
Melting Point 210°C
Color Gray-White to White
Physical Form Powder, Irregular Flakes or Granules
Quantity 500 g
Assay Percent Range 95%
Molecular Formula H2NNa
Linear Formula NaNH2
Fieser 01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288
Merck Index 15,8714
Solubility Information Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol
Molecular Weight (g/mol) 39
Formula Weight 39
Percent Purity 95%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Sodium amide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.