missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenate Dibasic Heptahydrate, ACS Reagent, 98.0 to 102.0%, Honeywell™ Fluka™

Product Code. 15628160 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15628160 50 g Poly bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15628160 Supplier Honeywell-Fluka Supplier No. S966350G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 10048-95-0
Quantity 50 g
Molecular Formula AsH15Na2O11
Linear Formula Na2HAsO4 · 7H2O
MDL Number MFCD00149179
UN Number NONH for all modes of transport
Synonym disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate
InChI Key KOLXPEJIBITWIQ-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O
IUPAC Name disodium;hydrogen arsorate;heptahydrate
Molecular Weight (g/mol) 312.01
PubChem CID 61460
Formula Weight 312.01g/mol
Percent Purity 98.0 to 102.0%
Packaging Poly bottle
Chemical Name or Material Sodium arsenate dibasic heptahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.