missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenate Heptahydrate, TRC

Product Code. 30412126
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412126 10 g 10g
30419003 5 g 5g
30428482 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412126 Supplier Toronto Research Chemicals Supplier No. TRCS96008810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10048-95-0
Molecular Formula HO4As2 Na • 7[H2O]
Molecular Weight (g/mol) 185.9171802
Synonym Dibasic Sodium Arsenate (Na2HAsO4.7H2O), Disodium Acid Arsenate Heptahydrate (Na2HAsO4.7H2O), Disodium Arsenate Heptahydrate, Disodium Hydrogen Arsenate (Na2HAsO4) Heptahydrate, Disodium Monohydrogen Arsenate Heptahydrate, Hydrogen Disodium Arsenate (HNa2AsO4) Heptahydrate, Sodium Acid Arsenate, Heptahydrate, Sodium Arsenate (Na2HAsO4) Heptahydrate, Sodium Arsenate Hydrate (Na2HAsO4.7H2O), Sodium Hydrogen Arsenate (Na2HAsO4) Heptahydrate
IUPAC Name disodium;hydrogen arsorate;heptahydrate
SMILES O.O.O.O.O.O.O.O[As](=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 10 g
InChI Formula InChI=1S/AsH3O4.2Na.7H2O/c2-1(3,4)5;;;;;;;;;/h(H3,2,3,4,5);;;7*1H2/q;2*+1;;;;;;;/p-2
Formula Weight 311.9626
Chemical Name or Material Sodium Arsenate Heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.