missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ascorbate, TRC

Product Code. 17968114
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17968114 10 g 10g
17900005 100 g 100g
17917264 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17968114 Supplier Toronto Research Chemicals Supplier No. TRCS61220010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6 H7 O6 . Na
Molecular Weight (g/mol) 198.11
Synonym Ascorbic acid, sodium deriv. (6CI,7CI), L-Ascorbic acid, monosodium salt (8CI,9CI), 3-Oxo-L-gulofuranolactone sodium, ASK-P 10KR, Ascorbic acid sodium salt, Ascorbicin, Ascorbin, CK 40, CK 40 (ascorbate), Cebitate, Cenolate, E 301, E 301 (antioxidant), HBL 508, Iskia C, L-Ascorbic acid sodium salt, Limcee, Monosodium L-ascorbate, Monosodium ascorbate, Natrascorb, Natri-C, Sodascorbate, Sodium (+)-L-ascorbate, Sodium 2-(1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Sodium L-(+)-ascorbate, Sodium L-ascorbate, Sodium ascorbate, Vitamin C Na, Vitamin C Na salt, Vitamin C sodium
IUPAC Name sodium;(2R)-2-[(1S)-1,2-dihydroxyethyl]-4-hydroxy-5-oxo-2H-furan-3-olate
SMILES [Na+].OC[C@H](O)[C@H]1OC(=O)C(=C1[O-])O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H8O6.Na/c7-1-2(8)5-3(9)4(10)6(11)12-5;/h2,5,7-10H,1H2;/q;+1/p-1/t2-,5+;/m0./s1
Formula Weight 198.014
Percent Purity >95%
Chemical Name or Material Ascorbic acid sodium salt (Vitamin C sodium salt)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.