missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, ≥99%, (Crystalline), MP Biomedicals

Product Code. 11476272 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476272 500 g 500g
11456272 25 g 25g
11466272 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476272 Supplier MP Biomedicals Supplier No. 0210289190
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26628-22-8 | Formula: N3Na | Weight: 65.011 g/mol

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Physical Form Crystalline
Quantity 500 g
Assay Percent Range ≥99%
Formula Weight 65
Chemical Name or Material Sodium Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.