missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, ≥99%

Product Code. 15895328 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895328 1 kg Plastic bottle 1kg
15835188 100 g Plastic bottle 100g
15845188 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15895328 Supplier Thermo Scientific Supplier No. J21610.A1
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Afficher la politique du retour.

CAS: 26628-22-8 ∣ N3Na ∣ 65.011 g/mol

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Spécification

Melting Point ∼275°C (decomposition)
Physical Form Crystalline Powder
Quantity 1 kg
Packaging Plastic bottle
Specific Gravity 1.846
Density 1.846
UN Number UN1687
Formula Weight 65.01
Percent Purity >99%
Grade Ungraded
Chemical Name or Material Sodium azide
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Sodium azide, Thermo Scientific
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Mention de danger
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Informations supplémentaires
  • MIXTURE LIST-Contain: Sodium azide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.