missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, Extra Pure, SLR, Fisher Chemical™

Product Code. 10325720 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10325720 500 g 500g
10776301 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10325720 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' S/2360/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N3Na, CAS Number-26628-22-8, azoture de sodium, hydrazoic acid, sodium salt, azide, sodium, sodium azide, kazoe, smite, nemazyd, natriumazid german, sodium azide na n3, natriumazid, 100g, 300 deg.C, CHEBI:278547, White, 65.01g/mol, PXIPVTKHYLBLMZ-UHFFFAOYSA-N, sodium;azide, 275 deg.C, 3536, 65.011

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number 3536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Boiling Point 300°C
Melting Point 275°C
Color White
pH 10
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 65.01g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.