missing translation for 'onlineSavingsMsg'
Learn More

Sodium azide, Honeywell™

Product Code. 16466349 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
Grade:
Reagent
Unit Size:
1kg
Product Code. Quantity Packaging Grade unitSize
16466349 1 kg Plastic Bottle Reagent 1kg
1 options
This item is not returnable. View return policy

Product Code. 16466349

Brand: Honeywell Research Chemicals 13412H1KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 26628-22-8
Color Colorless
pH >7.0 (20°C)
Quantity 1 kg
Molecular Formula N3Na
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES [Na+].[N-]=[N+]=[N-]
IUPAC Name sodium azide
Molecular Weight (g/mol) 65.01
Percent Purity 99%
Grade Reagent
Packaging Plastic Bottle
Chemical Name or Material Sodium azide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.