missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide OR, Macron Fine Chemicals™

Product Code. 15548674
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15548674 500 g Poly 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548674 Supplier Avantor Macron Supplier No. 195359
  • ONLINE EXCLUSIVE PRICE
    235.37 €
    206.16 € / 500g
    Save 29.21 €
    12% Off
 Restricted Item
Estimated Shipment: 07-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 65.01
Percent Purity ≥99.7%
Grade Not Available
Packaging Poly
Chemical Name or Material Sodium Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.