missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium benzoate, 99%, for biochemistry

Product Code. 12685107 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12685107 250 g 250g
12675107 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12685107 Supplier Thermo Scientific Chemicals Supplier No. 447802500
  • ONLINE EXCLUSIVE PRICE
    39.15 €
    36.70 € / 250g
    Save 2.45 €
    6% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
PubChem CID 517055
IUPAC Name sodium;benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CO2Na
Quantity 250 g
Merck Index 15,8718
Solubility Information (1 M in water) Clear colorless to light yellow
Formula Weight 144.11
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material Sodium benzoate
Show More Show Less
compliance-icons
Product Identifier
  • Benzoic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.