missing translation for 'onlineSavingsMsg'
Learn More

Sodium benzoate, 99%

Product Code. 11448496
Change view
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Unit Size:
10000g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448496 10,000 g 10000g
11433980 250 g 250g
11428496 500 g 500g
11438496 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11448496 Supplier Thermo Scientific Alfa Aesar Supplier No. A15946.0C
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium benzoate is widely used as a preservative in food, medicine, cosmetics and animal feeds. It is used in the treatment of hyperammonemia and urea cycle disorders. It is used in the fireworks as a fuel in whistle mix. It is also used in the preparation of toothpaste and mouthwashes. It finds application in most of the acidic foods such as salad dressings (vinegar), carbonated drinks (carbonic acid), jams and fruit juices (citric acid), pickles (vinegar), and condiments.

Solubility
Soluble in water, alcoholSoluble in water, alcohol, ammonia and pyridine.

Notes
Hygroscopic. Incompatible with alkalis, mineral acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
PubChem CID 517055
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point >300°C
Assay Percent Range 99%
Odor Odorless
Quantity 10,000 g
Beilstein 3572467
Merck Index 14,8582
Solubility Information Soluble in water,alcoholSoluble in water,alcohol,ammonia and pyridine.
IUPAC Name sodium;benzoate
Formula Weight 144.11
Percent Purity 99%
Physical Form Powder or granules or prills
Chemical Name or Material Sodium benzoate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium benzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.