missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Certified AR for Analysis, Fisher Chemical

Product Code. 10132210 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10132210 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10132210 Supplier Fisher Chemical Supplier No. S/2441/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 532-32-1
Color White
pH 8
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C7H5NaO2
MDL Number MFCD00012463
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1
IUPAC Name sodium benzoate
Molecular Weight (g/mol) 144.11
PubChem CID 517055
Formula Weight 144.11g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Benzoic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.