missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10488990 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10488990 1 kg 1kg
10284060 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10488990 Supplier Fisher Chemical Supplier No. S/2440/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7H5NaO2
Molecular Weight (g/mol) 144.11
MDL Number MFCD00012463
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
PubChem CID 517055
IUPAC Name sodium;benzoate
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point 300°C
Color White
pH 8
Physical Form Powder/Solid
Quantity 1 kg
Formula Weight 144.11g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Benzoic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.