missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15558134 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15558134 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558134 Supplier Avantor J.T.Baker Supplier No. 350105
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multi-compendial grade sodium benzoate for manufacturing, processing, or repackaging applications.

Specifications

CAS 532-32-1
Physical Form Powder
Quantity 2.5 kg
Molecular Formula C7H5NaO2
MDL Number MFCD00012463
Synonym sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746
InChI Key WXMKPNITSTVMEF-UHFFFAOYSA-M
SMILES [Na+].[O-]C(=O)C1=CC=CC=C1
IUPAC Name sodium benzoate
Molecular Weight (g/mol) 144.11
PubChem CID 517055
Formula Weight 144.11
Percent Purity 1.006
Grade Multi-Compendial/NF
Packaging Poly Bottle
Specific Gravity 1L = 1.44kg
Chemical Name or Material Sodium Benzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.