missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, TRC

Product Code. 30008970
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008970 500 g 500g
30007179 100 g 100g
17911685 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008970 Supplier Toronto Research Chemicals Supplier No. TRCS614000500G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-32-1
Molecular Formula C7 H5 O2 . Na
Molecular Weight (g/mol) 144.1
Synonym Benzoic acid, sodium salt (8CI,9CI), Sodium benzoate (6CI), Antimol, Benzoate of soda, E 211, E 221, E 221 (nucleating agent), Fuminaru, Microcare SB, Probenz, Purox S, Sobenate
IUPAC Name sodium;benzoate
SMILES [Na+].[O-]C(=O)c1ccccc1

Specifications

Quantity 500 g
InChI Formula InChI=1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1
Formula Weight 144.0187
Percent Purity >95%
Chemical Name or Material Benzoic Acid Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.