missing translation for 'onlineSavingsMsg'
Learn More

Sodium beta-glycerophosphate pentahydrate, 97%

Product Code. 11312156
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312156 100 g 100g
11302156 25 g 25g
11322156 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11312156 Supplier Thermo Scientific Alfa Aesar Supplier No. L03425.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13408-09-8 | C3H19Na2O11P | 308.13 g/mol

Sodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium β-glycerophosphate pentahydrate is used as a phosphatase inhibitor. It promotes bone matrix mineralization while delivering to osteoblasts by providing a source of phosphate ions. It is used in the development of hydrogels and scaffolds, which finds applications in tissue engineering and cell growth. It is used as an additive in isolation mediums by providing phosphate ions to isolate. It is utilized to promote mineralization in vitro by modulating bone cell metabolic activity.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13408-09-8
Molecular Formula C3H19Na2O11P
Molecular Weight (g/mol) 308.13
MDL Number MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
InChI Key VWVLGHQKEJESPW-UHFFFAOYSA-N
Synonym unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
PubChem CID 131846916
IUPAC Name 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate
SMILES O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O

Specifications

Melting Point >300°C
pH 9.5
Quantity 100 g
Beilstein 3744328
Merck Index 14,8622
Solubility Information Soluble in water.
Formula Weight 306.12 (216.04 Anhydrous)
Percent Purity 97%
Chemical Name or Material Sodium beta-glycerophosphate pentahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.