missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium bicarbonate, 99.5%, for biochemistry

Product Code. 12655057 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12655057 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12655057 Supplier Thermo Scientific Chemicals Supplier No. 447100010
  • ONLINE EXCLUSIVE PRICE
    70.90 €
    68.80 € / 1kg
    Save 2.10 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144-55-8 ∣ CHNaO3 ∣ 84.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 270.0°C
CAS 144-55-8
Physical Form Crystalline Powder
Quantity 1 kg
Packaging Plastic Bottle
Molecular Formula CHNaO3
Linear Formula NaHCO3
MDL Number MFCD00003528
Merck Index 158719
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
Solubility Information Clear colorless
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES [Na+].OC([O-])=O
IUPAC Name sodium;hydrogen carbonate
Molecular Weight (g/mol) 84.01
PubChem CID 516892
ChEBI CHEBI:32139
Formula Weight 84.01
Percent Purity 99.5%
Grade Biochemical
Chemical Name or Material Sodium bicarbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.