missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, 99+%, Extra Pure

Product Code. 10694782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694782 1 kg Plastic bottle 1kg
10051210 5 kg Plastic drum 5kg
10493635 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10694782 Supplier Thermo Scientific Acros Supplier No. 123360010
  • ONLINE EXCLUSIVE PRICE
    32.15 €
    30.10 € / 1kg
    Save 2.05 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium bicarbonate, 99+%, CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 5kg, 20ppm max., 100ppm max., CHEBI:32139, White, 205-633-8 | CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 1 kg
Packaging Plastic bottle
Assay Percent Range 99+%
Linear Formula NaHCO3
Fieser 03,260; 05,595; 11,476; 17,180
Merck Index 15, 8719
Solubility Information Solubility in water: 9g/100mL (20°C). Other solubilities: 16.4g/100mL water (60°C),insoluble in alcohol
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Sodium bicarbonate
Show More Show Less

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.