missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate for HPLC, ≥99.0%, Honeywell™ Fluka™

Product Code. 15657370 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 15657370

Brand: Honeywell-Fluka 88208250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for HPLC,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 144-55-8
Quantity 250 g
Molecular Formula CHNaO3
Linear Formula NaHCO3
MDL Number MFCD00003528
UN Number NONH for all modes of transport
Beilstein 4153970
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES [Na+].OC([O-])=O
IUPAC Name sodium;hydrogen carbonate
Molecular Weight (g/mol) 84.01
PubChem CID 516892
ChEBI CHEBI:32139
Formula Weight 84.01g/mol
Percent Purity ≥99.0%
Packaging Glass bottle
Chemical Name or Material Sodium bicarbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.